2-Butoxy-3-chloroaniline

Supplier: Matrix Scientific

021878-500MG
101812-918EA 235.75 USD
101812-918
2-Butoxy-3-chloroaniline
2-Butoxy-3-chloroaniline

Hazard: Irritant

Formula: C₁₀H₁₄ClNO
MDL Number: MFCD08687148
CAS Number: 130566-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR