2-(2-Naphthyloxy)aniline

Supplier: Matrix Scientific

021865-500MG
101812-894EA 315.38 USD
101812-894
2-(2-Naphthyloxy)aniline
2-(2-Naphthyloxy)aniline

Hazard: Irritant

Formula: C₁₆H₁₃NO
MDL Number: MFCD00506410
CAS Number: 32219-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR