2-[(4-Chloro-1-naphthyl)oxy]aniline

Supplier: Matrix Scientific

021864-500MG
101812-892EA 315.38 USD
101812-892
2-[(4-Chloro-1-naphthyl)oxy]aniline
2-[(4-Chloro-1-naphthyl)oxy]aniline

Hazard: Irritant

Formula: C₁₆H₁₂ClNO
MDL Number: MFCD08687403
CAS Number: 946727-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR