2-(2-Methoxy-4-methylphenoxy)phenylamine
Supplier: Matrix Scientific
021858-500MG
101812-880EA
315.38
USD
101812-880
2-(2-Methoxy-4-methylphenoxy)phenylamine
2-(2-Methoxy-4-methylphenoxy)phenylamine
Hazard: Irritant
Formula:
C₁₄H₁₅NO₂ |
MDL Number:
MFCD08688054 CAS Number: 937607-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...