2-(4-Methylphenoxy)aniline

Supplier: Matrix Scientific

021857-500MG
101812-878EA 315.38 USD
101812-878
2-(4-Methylphenoxy)aniline
2-(4-Methylphenoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₃NO
MDL Number: MFCD00035923
CAS Number: 20927-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR