2-(4-Isopropylphenoxy)aniline

Supplier: Matrix Scientific

021856-500MG
101812-876EA 315.38 USD
101812-876
2-(4-Isopropylphenoxy)aniline
2-(4-Isopropylphenoxy)aniline

Hazard: Irritant

Formula: C₁₅H₁₇NO
MDL Number: MFCD08687983
CAS Number: 640766-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR