Methyl-3-(2-aminophenoxy)benzoate

Supplier: Matrix Scientific

021839-500MG
101812-842EA 235.75 USD
101812-842
Methyl-3-(2-aminophenoxy)benzoate
Methyl-3-(2-aminophenoxy)benzoate

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
MDL Number: MFCD08688146
CAS Number: 227275-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR