2-[3-(Dimethylamino)phenoxy]aniline

Supplier: Matrix Scientific

021837-500MG
101812-838EA 235.75 USD
101812-838
2-[3-(Dimethylamino)phenoxy]aniline
2-[3-(Dimethylamino)phenoxy]aniline

Hazard: Irritant

Formula: C₁₄H₁₆N₂O
MDL Number: MFCD08687609
CAS Number: 349667-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR