N-[3-(2-Aminophenoxy)phenyl]acetamide hydrochloride

Supplier: Matrix Scientific

021836-500MG
101812-836EA 139.15 USD
101812-836
N-[3-(2-Aminophenoxy)phenyl]acetamide hydrochloride
N-[3-(2-Aminophenoxy)phenyl]acetamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₅ClN₂O₂
MDL Number: MFCD11506372
CAS Number: 1185304-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR