2-(3-Chloro-4-fluorophenoxy)aniline

Supplier: Matrix Scientific

021835-500MG
101812-834EA 315.38 USD
101812-834
2-(3-Chloro-4-fluorophenoxy)aniline
2-(3-Chloro-4-fluorophenoxy)aniline

Hazard: Irritant

Formula: C₁₂H₉ClFNO
MDL Number: MFCD08687307
CAS Number: 946714-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR