2-(3-Chlorophenoxy)aniline
Supplier: Matrix Scientific
021834-500MG
101812-832EA
315.38
USD
101812-832
2-(3-Chlorophenoxy)aniline
2-(3-Chlorophenoxy)aniline
Hazard: Irritant
Formula:
C₁₂H₁₀ClNO |
MDL Number:
MFCD04970990 CAS Number: 76838-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...