2-(2-Ethylphenoxy)aniline

Supplier: Matrix Scientific

021832-500MG
101812-828EA 315.38 USD
101812-828
2-(2-Ethylphenoxy)aniline
2-(2-Ethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₅NO
MDL Number: MFCD08687763
CAS Number: 4475-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR