2-(2-Fluorophenoxy)aniline

Supplier: Matrix Scientific

021819-500MG
101812-802EA 315.38 USD
101812-802
2-(2-Fluorophenoxy)aniline
2-(2-Fluorophenoxy)aniline

Hazard: Irritant

Formula: C₁₂H₁₀FNO
MDL Number: MFCD04970974
CAS Number: 391906-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR