2-Phenoxyaniline

Supplier: Matrix Scientific

Synonyms: 1-Amino-2-phenoxybenzol

021818-500MG
101812-800EA 235.75 USD
101812-800
2-Phenoxyaniline
2-Phenoxyaniline

Hazard: Irritant

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
MDL Number: MFCD00035765
CAS Number: 2688-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR