2-(3-Methoxypropoxy)aniline

Supplier: Matrix Scientific

021816-500MG
101812-796EA 315.38 USD
101812-796
2-(3-Methoxypropoxy)aniline
2-(3-Methoxypropoxy)aniline

Hazard: Irritant

Formula: C₁₀H₁₅NO₂
MDL Number: MFCD08688105
CAS Number: 857272-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR