2-[2-(Methylanilino)ethoxy]aniline
Supplier: Matrix Scientific
021815-500MG
101812-794EA
315.38
USD
101812-794
2-[2-(Methylanilino)ethoxy]aniline
2-[2-(Methylanilino)ethoxy]aniline
Hazard: Irritant
Formula:
C₁₅H₁₈N₂O |
MDL Number:
MFCD08688160 CAS Number: 946772-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...