2-(Cyclohexylmethoxy)aniline

Supplier: Matrix Scientific

021806-500MG
101812-778EA 235.75 USD
101812-778
2-(Cyclohexylmethoxy)aniline
2-(Cyclohexylmethoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08687494
CAS Number: 937607-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR