2-(Cyclopentylmethoxy)aniline

Supplier: Matrix Scientific

021805-500MG
101812-776EA 384.62 USD
101812-776
2-(Cyclopentylmethoxy)aniline
2-(Cyclopentylmethoxy)aniline

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD08687508
CAS Number: 640767-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR