3-(1-Piperidinyl)-8-azabicyclo[3.2.1]octane dihydrochloride

Supplier: Matrix Scientific

021799-500MG
101812-764EA 402.5 USD
101812-764
3-(1-Piperidinyl)-8-azabicyclo[3.2.1]octane dihydrochloride
3-(1-Piperidinyl)-8-azabicyclo[3.2.1]octane dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₄Cl₂N₂
MDL Number: MFCD11100474
CAS Number: 1185301-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR