N,N-Dimethyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride

Supplier: Matrix Scientific

021798-500MG
101812-762EA 315.38 USD
101812-762
N,N-Dimethyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride
N,N-Dimethyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂
MDL Number: MFCD09800619
CAS Number: 1185301-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR