(4-Fluoro-4'-methoxy[1,1'-biphenyl]-3-yl)methanamine

Supplier: Matrix Scientific

021795-500MG
101812-756EA 235.75 USD
101812-756
(4-Fluoro-4'-methoxy[1,1'-biphenyl]-3-yl)methanamine
(4-Fluoro-4'-methoxy[1,1'-biphenyl]-3-yl)methanamine

Hazard: Irritant

Formula: C₁₄H₁₄FNO
MDL Number: MFCD08687836
CAS Number: 946682-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR