(4'-Methoxy-2'-methyl[1,1'-biphenyl]-2-yl)methanamine

Supplier: Matrix Scientific

021787-500MG
101812-740EA 315.38 USD
101812-740
(4'-Methoxy-2'-methyl[1,1'-biphenyl]-2-yl)methanamine
(4'-Methoxy-2'-methyl[1,1'-biphenyl]-2-yl)methanamine

Hazard: Irritant

Formula: C₁₅H₁₇NO
MDL Number: MFCD08688049
CAS Number: 946726-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR