(2'-Fluoro[1,1'-biphenyl]-2-yl)methanamine

Supplier: Matrix Scientific

021783-500MG
101812-732EA 538.46 USD
101812-732
(2'-Fluoro[1,1'-biphenyl]-2-yl)methanamine
(2'-Fluoro[1,1'-biphenyl]-2-yl)methanamine

Hazard: Irritant

Formula: C₁₃H₁₂FN
MDL Number: MFCD05980874
CAS Number: 946714-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR