2'-(Aminomethyl)-N,N-dimethyl[1,1'-biphenyl]-4-amine

Supplier: Matrix Scientific

021782-500MG
101812-730EA 538.46 USD
101812-730
2'-(Aminomethyl)-N,N-dimethyl[1,1'-biphenyl]-4-amine
2'-(Aminomethyl)-N,N-dimethyl[1,1'-biphenyl]-4-amine

Hazard: Irritant

Formula: C₁₅H₁₈N₂
MDL Number: MFCD08686809
CAS Number: 946682-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR