(3'-Methoxy[1,1'-biphenyl]-2-yl)methanamine

Supplier: Matrix Scientific

021781-500MG
101812-728EA 538.46 USD
101812-728
(3'-Methoxy[1,1'-biphenyl]-2-yl)methanamine
(3'-Methoxy[1,1'-biphenyl]-2-yl)methanamine

Hazard: Irritant

Formula: C₁₄H₁₅NO
MDL Number: MFCD08688023
CAS Number: 946726-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR