(2'-Methoxy[1,1'-biphenyl]-2-yl)methanamine

Supplier: Matrix Scientific

021778-500MG
101812-722EA 402.5 USD
101812-722
(2'-Methoxy[1,1'-biphenyl]-2-yl)methanamine
(2'-Methoxy[1,1'-biphenyl]-2-yl)methanamine

Hazard: Irritant

Formula: C₁₄H₁₅NO
MDL Number: MFCD08688022
CAS Number: 292151-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR