2-(4-Methoxy-3-methylphenyl)azepane

Supplier: Matrix Scientific

021776-500MG
101812-720EA 538.46 USD
101812-720
2-(4-Methoxy-3-methylphenyl)azepane
2-(4-Methoxy-3-methylphenyl)azepane

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD05190342
CAS Number: 887361-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR