2-(4-Methoxy-3,5-dimethylphenyl)azepane

Supplier: Matrix Scientific

021772-500MG
101812-712EA 315.38 USD
101812-712
2-(4-Methoxy-3,5-dimethylphenyl)azepane
2-(4-Methoxy-3,5-dimethylphenyl)azepane

Hazard: Irritant

Formula: C₁₅H₂₃NO
MDL Number: MFCD05190365
CAS Number: 901921-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR