4-(4-Methoxy-2-methylphenyl)piperidine hydrochloride

Supplier: Matrix Scientific

021769-500MG
101812-706EA 235.75 USD
101812-706
4-(4-Methoxy-2-methylphenyl)piperidine hydrochloride
4-(4-Methoxy-2-methylphenyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD09997050
CAS Number: 1185297-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR