4-(2,4-Dimethoxyphenyl)piperidine hydrochloride

Supplier: Matrix Scientific

021763-500MG
101812-694EA 402.5 USD
101812-694
4-(2,4-Dimethoxyphenyl)piperidine hydrochloride
4-(2,4-Dimethoxyphenyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD10687207
CAS Number: 1185297-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR