2-(3,4-Dimethoxyphenyl)azepane
Supplier: Matrix Scientific
021762-500MG
101812-692EA
315.38
USD
101812-692
2-(3,4-Dimethoxyphenyl)azepane
2-(3,4-Dimethoxyphenyl)azepane
Hazard: Irritant
Formula:
C₁₄H₂₁NO₂ |
MDL Number:
MFCD02663724 CAS Number: 32084-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...