4-(3,4-Dimethoxyphenyl)piperidine hydrochloride

Supplier: Matrix Scientific

021761-500MG
101812-690EA 538.46 USD
101812-690
4-(3,4-Dimethoxyphenyl)piperidine hydrochloride
4-(3,4-Dimethoxyphenyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD08751545
CAS Number: 121278-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR