2-[3-(Trifluoromethyl)phenyl]azepane

Supplier: Matrix Scientific

021758-500MG
101812-686EA 315.38 USD
101812-686
2-[3-(Trifluoromethyl)phenyl]azepane
2-[3-(Trifluoromethyl)phenyl]azepane

Hazard: Irritant

Formula: C₁₃H₁₆F₃N
MDL Number: MFCD02663827
CAS Number: 383131-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR