2-(4-Fluorophenyl)azepane

Supplier: Matrix Scientific

021756-500MG
101812-682EA 235.75 USD
101812-682
2-(4-Fluorophenyl)azepane
2-(4-Fluorophenyl)azepane

Hazard: Irritant

Formula: C₁₂H₁₆FN
MDL Number: MFCD02663706
CAS Number: 168890-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR