2-(2-Fluorophenyl)azepane

Supplier: Matrix Scientific

021750-500MG
101812-670EA 454.25 USD
101812-670
2-(2-Fluorophenyl)azepane
2-(2-Fluorophenyl)azepane

Hazard: Irritant

Formula: C₁₂H₁₆FN
MDL Number: MFCD02663777
CAS Number: 383130-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR