2-(4-Methylphenyl)azepane

Supplier: Matrix Scientific

021748-500MG
101812-666EA 235.75 USD
101812-666
2-(4-Methylphenyl)azepane
2-(4-Methylphenyl)azepane

Hazard: Irritant

Formula: C₁₃H₁₉N
MDL Number: MFCD02663755
CAS Number: 168890-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR