4-(4-Methylphenyl)piperidine hydrochloride

Supplier: Matrix Scientific

021747-500MG
101812-664EA 538.46 USD
101812-664
4-(4-Methylphenyl)piperidine hydrochloride
4-(4-Methylphenyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClN
MDL Number: MFCD02178906
CAS Number: 67765-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR