4-[4-(tert-Butyl)phenyl]piperidine hydrochloride

Supplier: Matrix Scientific

021745-500MG
101812-660EA 315.38 USD
101812-660
4-[4-(tert-Butyl)phenyl]piperidine hydrochloride
4-[4-(tert-Butyl)phenyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClN
MDL Number: MFCD11100472
CAS Number: 1185297-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR