4-(4-Ethoxyphenyl)piperidine hydrochloride

Supplier: Matrix Scientific

021743-500MG
101812-656EA 315.38 USD
101812-656
4-(4-Ethoxyphenyl)piperidine hydrochloride
4-(4-Ethoxyphenyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD11845766
CAS Number: 170856-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR