2-(3,4-Dimethylphenyl)azepane

Supplier: Matrix Scientific

021735-500MG
101812-644EA 495.38 USD
101812-644
2-(3,4-Dimethylphenyl)azepane
2-(3,4-Dimethylphenyl)azepane

Hazard: Irritant

Formula: C₁₄H₂₁N
MDL Number: MFCD05190468
CAS Number: 901924-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR