4-(3-Methylphenyl)piperidine hydrochloride

Supplier: Matrix Scientific

021732-500MG
101812-638EA 495.38 USD
101812-638
4-(3-Methylphenyl)piperidine hydrochloride
4-(3-Methylphenyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClN
MDL Number: MFCD02178907
CAS Number: 80120-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR