2-(2,4,5-Trimethylphenyl)azepane

Supplier: Matrix Scientific

021728-500MG
101812-630EA 235.75 USD
101812-630
2-(2,4,5-Trimethylphenyl)azepane
2-(2,4,5-Trimethylphenyl)azepane

Hazard: Irritant

Formula: C₁₅H₂₃N
MDL Number: MFCD08688566
CAS Number: 946714-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR