4-(2-Methoxyphenyl)piperidine hydrochloride
Supplier: Matrix Scientific
021723-500MG
101812-620EA
315.38
USD
101812-620
4-(2-Methoxyphenyl)piperidine hydrochloride
4-(2-Methoxyphenyl)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClNO |
MDL Number:
MFCD02178920 CAS Number: 82212-04-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...