3,3'-Difluoro[1,1'-biphenyl]-4-ylamine hydrochloride

Supplier: Matrix Scientific

021717-500MG
101812-608EA 495.38 USD
101812-608
3,3'-Difluoro[1,1'-biphenyl]-4-ylamine hydrochloride
3,3'-Difluoro[1,1'-biphenyl]-4-ylamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₀ClF₂N
MDL Number: MFCD11506370
CAS Number: 1185296-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR