N-(4'-Amino-3'-fluoro[1,1'-biphenyl]-4-yl)-N,N-dimethylamine

Supplier: Matrix Scientific

021715-500MG
101812-604EA 315.38 USD
101812-604
N-(4'-Amino-3'-fluoro[1,1'-biphenyl]-4-yl)-N,N-dimethylamine
N-(4'-Amino-3'-fluoro[1,1'-biphenyl]-4-yl)-N,N-dimethylamine

Hazard: Irritant

Formula: C₁₄H₁₅FN₂
MDL Number: MFCD08686795
CAS Number: 946681-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR