3-Fluoro-3',4'-dimethyl[1,1'-biphenyl]-4-ylamine

Supplier: Matrix Scientific

021714-500MG
101812-602EA 315.38 USD
101812-602
3-Fluoro-3',4'-dimethyl[1,1'-biphenyl]-4-ylamine
3-Fluoro-3',4'-dimethyl[1,1'-biphenyl]-4-ylamine

Hazard: Irritant

Formula: C₁₄H₁₄FN
MDL Number: MFCD08687812
CAS Number: 946713-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR