3-Fluoro-2'-methyl[1,1'-biphenyl]-4-amine

Supplier: Matrix Scientific

021711-500MG
101812-596EA 235.75 USD
101812-596
3-Fluoro-2'-methyl[1,1'-biphenyl]-4-amine
3-Fluoro-2'-methyl[1,1'-biphenyl]-4-amine

Hazard: Irritant

Formula: C₁₃H₁₂FN
MDL Number: MFCD06801813
CAS Number: 893735-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR