3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-amine

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4'-methoxy-[1,1'-biphenyl]-4-amine

021710-500MG
101812-594EA 315.38 USD
101812-594
3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-amine
3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-amine

Hazard: Irritant

Formula: C₁₃H₁₂FNO
MW: 217.25 g/mol
MDL Number: MFCD06802177
CAS Number: 893734-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR