3'-(Trifluoromethyl)[1,1'-biphenyl]-3-amine

Supplier: Matrix Scientific

Synonyms: 3'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine

021706-500MG
101812-586EA 254.15 USD
101812-586
3'-(Trifluoromethyl)[1,1'-biphenyl]-3-amine
3'-(Trifluoromethyl)[1,1'-biphenyl]-3-amine

Hazard: Irritant

Formula: C₁₃H₁₀F₃N
MW: 237.23 g/mol
MDL Number: MFCD03424670
CAS Number: 400749-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR