3',4'-Dimethyl[1,1'-biphenyl]-3-amine

Supplier: Matrix Scientific

021705-500MG
101812-584EA 235.75 USD
101812-584
3',4'-Dimethyl[1,1'-biphenyl]-3-amine
3',4'-Dimethyl[1,1'-biphenyl]-3-amine

Hazard: Irritant

Formula: C₁₄H₁₅N
MDL Number: MFCD04117436
CAS Number: 728919-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR